KIMBLE® PTFE-Faced Red Rubber Septa, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
73816-15 73816-18 73816-20 73816-24 N73812-13
66010-720CS 186.22 CAD
66010-720 66010-730 66010-741 66010-912 93001-092 93001-094
KIMBLE® PTFE-Faced Red Rubber Septa, DWK Lifesciences
Septa
Highly chemical-resistant PTFE facing maintains integrity of contents.
  • Economical and excellent resealability
  • Low extractables
  • Resists coring
  • PTFE thickness: 0.002"