SatPax® 550 Pre-Wetted 100% Meltblown Polypropylene Cleanroom Wiper, Berkshire
Supplier: Berkshire
SPX550.00124 SPX55000424 SPX550.005.36 SPX550.003.36 SPX55000836
89138-004CS 1211.83 CAD
89138-004 75831-354 76186-222 76196-332 76638-268
SatPax® 550 Pre-Wetted 100% Meltblown Polypropylene Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SatPax® 550 wipers are exceptionally clean. This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents, maintenance of squirt bottles, and inconsistent wetting and cleaning associated with wetting a dry wiper.
  • 100% meltblown polypropylene
  • Pre-wetted with 70% IPA/30% DI water
  • Re-sealable solvent resistant packaging
  • Knife cut edges
  • Excellent wet strength and abrasion resistance
  • Reduces alcohol usage and preparation/handling cost
  • Increases cleaning efficiency
SatPax® 550 wipers feature very low extractables and ions, as well as low particles and fibre counts.
Designed for use in ISO Class 5 and higher cleanroom environments and USP <797> applications.
These wipers are composed of exceptionally clean 100% meltblown polypropylene, pre-wetted with 70% IPA and 30% DI water. This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents.