MicroPolx® 1100 Sealed Edge Cleanroom Wiper, Berkshire
Supplier: Berkshire
MP1100090914 MP1100121210
75801-080CS 1023.14 CAD
75801-080 75801-082
MicroPolx® 1100 Sealed Edge Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
MicroPolx® 1100 is a 100% polyester microfiber sealed edge cleanroom wiper.
  • Stable double knit construction and high fiber surface area for improved wiping efficiency
  • Soft and smooth surface is suitable for cleaning sensitive or scratch prone surfaces
  • Chemically compatible with IPA and other commonly used solvents
Laser cut edges and a proprietary laundering process provide critically low particle and fiber release, as well as best-in-class ion and non-volatile residue levels.
The wiper’s microfiber yarn has three times the surface area of standardMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable