LIBERO CD Dry Ice Data Logger, ELPRO
Supplier: ELPRO SERVICES, INC.
802065
75870-732BX 5616 CAD
75870-732
LIBERO CD Dry Ice Data Logger, ELPRO
Data Loggers
ELPRO's LIBERO CD dry ice data logger provides the most accurate read on cold chain packing performance before and during shipments. Designed to measure even the lowest temperatures seen occurring in dry ice shipments at -95 °C.
  • Internal sensor works directly exposed to dry ice
  • Concise reporting in one file: ELPRO’s patented one-page PDF reporting includes embedded raw data, eliminating the hassle of multiple files
  • Multi-level alarm with eight ranges plus shipment duration as alarm criteria, customer configurabl, no need to pre-order profiles
  • Auto Expiry Date Management: Expiry date printed on every logger and programmed; virtually eliminates the risk of using an expired logger
  • Button cell battery avoids customs delays and dangerous goods label requirements for lithium battery air shipments; IATA compliant
  • No software required due to easy USB connection allows for easy data downloading
Users are able to save valuable time during shipments and start monitoring or downloading their data immediately due to a programmable site delay; avoiding time-consuming pre-conditioning/equilibrating of the device. Ideal for dry ice, transport.
The LIBERO CD is fully compatible with ELPRO’s monitoring software solutions including liberoMANAGER (cold chain cloud database); liberoCONFIG (configuration); elproVIEWER (analysis) and elproAssistant.
Certifications: IP 65, Fully calibrated with traceable certificate; GAMP5 validated.