Vanadium(III) oxide ≥99.7%, powder

Supplier: Thermo Scientific Chemicals
Danger

43377-22 43377-36
CAAA43377-22EA 486.64 CAD
CAAA43377-22 CAAA43377-36
Vanadium(III) oxide ≥99.7%, powder
Vanadium (III) oxide
Formula: O₃V₂
MW: 149.88
Melting Pt: 1940
Density: 4.87
Storage Temperature: Ambient
MDL Number: MFCD00011455
CAS Number: 1314-34-7
EINECS: 215-230-9
UN: 3285
ADR: 6.1 III
Merck Index: 14,9923

Order Now

Specification Test Results

Total Metal Impurities 0.3% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR