EB Buffer, sterile, Teknova
Supplier: TEKNOVA CA
E2010
CA100218-580EA 790.47 CAD
CA100218-580
EB Buffer, sterile, Teknova
Buffers Biological Buffers
EB Buffer, HEPES 20 mM, NaCl 420 mM, KCl 10 mM, MgCl2 1mM, Glycerol 20%, 0.1% Triton X-100, pH 7.0
Order Now
SPECIFICATIONS
Environmentally Preferable