Silicic acid calcium salt NF

Supplier: Spectrum Chemical Mfg. Corp.

C1643-100GM C1643-1KG C1643-500GM
75810-848EA 242.29 CAD
75810-848 75810-850 75810-852
Silicic acid calcium salt NF
Silicic acid calcium salt

Calcium Silicate, NF, also known as calcium orthosilicate, is used as an anticaking agent in pharmaceutical preparations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Certifications: NF product are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
CAS Number: 1344-95-2

Order Now

Specification Test Results

Calcium oxide (CaO) 20 - 35%
Silicon Dioxide (SiO₂) 50 - 65%
Sum of Calcium oxide, silicon dioxideand loss on ignition 90.0% Min.
pH (1 to 20 suspension) 8.4 - 11.2
Ratio of silicon dioxide to calcium oxide 0.5 - 20
Loss on ignition 20.0% Max.
Lead (Pb) 10 ppm Max.
Fluoride 50 ppm Max.
Identification Passes test
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR