BioMasher® II Micro Tissue Homogenizers, Closed System, Disposable, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
749625-0010 749625-0030 749625-0020
KT749625-0010CS 253.52 CAD
KT749625-0010 KT749625-0030 KT749625-0020
BioMasher® II Micro Tissue Homogenizers, Closed System, Disposable, Kimble Chase
Homogenizers Tissue Grinders
BioMasher® II is a micro sized closed system tissue homogenizers having a pestle with a molded-in shaft guard to minimize potential exposure to hazardous materials.
  • Clear polypropylene tube
  • Polyacetal pestle
The abrasive surfaces on the pestle tip and inner tube area ensure efficient grinding of samples. Ideal for homogenizing of animal tissues and organs as well as plant materials, insects, DNA, RNA, proteins, yeasts and enzymes.