F.I.T.™ Emergency Shower Station Mat Kits, Wearwell®
Supplier: Wearwell
546.58X27X30RDYL 546.58X27X42RDYL
75876-244EA 630.37 CAD
75876-244 75876-318
F.I.T.™ Emergency Shower Station Mat Kits, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
The Emergency Shower Mat Kit helps claim the space for any Emergency Shower Station.
  • Provides unencumbered access
  • Created for high visibility per ANSI recommendations
  • Elevates user above hazardous overflow
  • Conforms to ANSI Z535.1-1998 Standard
F.I.T.™ Kits are a remarkably simple, compact, and versatile matting solution for Emergency Showers. Mats feature high visibility yellow borders and elevate the user above hazardous overflow in critical situations. F.I.T.™ Kits for Emergency Shower Stations are a must for every facility.
Certifications: ASTMMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable