F.I.T.™ Emergency Eyewash Station Mat Kits, Wearwell®
Supplier: Wearwell
546.58X27X30GNYL 546.58X27X30RDYL
75876-242EA 635.43 CAD
75876-242 75876-246
F.I.T.™ Emergency Eyewash Station Mat Kits, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
The Emergency Eyewash Station Mat Kit helps claim the space for any Emergency Eyewash Station.
  • Provides unencumbered access
  • Created for high visibility per ANSI recommendations
  • Elevates user above hazardous overflow
  • Conforms to ANSI Z535.1-1998 Standard
F.I.T.™ Kits are a remarkably simple, compact, and versatile matting solution for Eyewash Stations. Mats feature high visibility yellow borders and elevate the user above hazardous overflow in critical situations. F.I.T.™ Kits for Emergency Eyewash Stations are a must for every facility.
Certifications: ASTM F1677. Dry: COF = 0.81. Wet: COF = 0.49.