F.I.T.™ Open Modular Interlocking Floor Tiles, Wearwell®
Supplier: Wearwell
540.58X12X12BK-CS20 540.58X3X12YL-CS20 540.58X3X15YL-CS4
75876-372CS 477.79 CAD
75876-372 75876-374 75876-376 75876-378 75876-380
F.I.T.™ Open Modular Interlocking Floor Tiles, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
F.I.T.™ is a versatile interlocking ergonomic flooring system.
  • Most versatile modular tile
  • Customizable in 3" increments
  • Secure and seamless connections
  • Ability to replace single tiles saves money and down time
  • Silicone-free
  • Ideally suited for oily environments
The most versatile interlocking ergonomic flooring system in the market. Patented LockSafe connections simply snap together and configure to fit an exact workspace. F.I.T.™ Open features an open-hole design that allows for rapid and complete fluid drainage. The surface slants towards the holes to facilitate quick drainage and to assure a safe, dry standing area for the worker. 100% silicone-free PVC tiles. Beveled ramps are available to finish the perimeter of any F.I.T.™ installation. Use F.I.T.™ Open to convert wet and dirty floor areas into ergonomic work zones.
Certifications: ASTM F1677. Dry: COF = 0.81. Wet: COF = 0.49.
Accessories information: The 3" wide safety edging works with all F.I.T.™ floor tiles. Add the edging for a finished look and for easy on/off access. Available in black and yellow.