Rejuvenator® Polyurethane Squared Surface Matting, Wearwell®
Supplier: Wearwell
502.58X2X5BK 502.58X2X3BK 502.58X3X5BK
75876-236EA 875.87 CAD
75876-236 75876-304 75876-310
Rejuvenator® Polyurethane Squared Surface Matting, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Rejuvenator® workstation mats provide superior anti-fatigue benefits and long-term performance.
  • 100% urethane with thick outer wear layer
  • Exceptional comfort and long-lasting performance
  • Molded-in safety bevels
  • Meets USP 797 compliance standards
  • Ideally suited for dry environments
Rejuvenator® WorkStation Mats provide an out-of-the-box anti-fatigue solution for workers who spend most of their shift standing at one station. Molded ramps on all four sides ensure safe on/off access. Same aesthetics as all other Rejuvenator® squared products, allowing any facility to look clean and consistent.
Certifications: ASTM F1677. Dry: COF = 0.75. Wet: COF = 0.28.