VWR® Flexible Multi-Tube Racks
Supplier: VWR International
HS120294 HS120295 HS120296 HS120297 HS120298 HS120299 HS120300 HS120293
75845-988CS 243.1 CAD
75845-988 75845-990 75845-992 75845-994 75845-996 75845-998 75846-000 75846-072
VWR® Flexible Multi-Tube Racks
Racks Tube Racks
This rack is truly adaptable, as each well adjusts to accommodate 7 to 50 mL tube sizes and holds them in a firm upright position.
  • Racks can be linked together for additional capacity
  • Open sides allow for label viewing
  • Flat base to hold tubes upright
  • Accommodates tubes from 7 to 50 mL sizes
  • Autoclavable
VWR® Flexible Multi-Tube Racks feature unique slots that adapt to accommodate varied tubes sizes and hold them in a firm upright position. Racks are manufactured from POM, and can be linked together for additional holding capacity. Linked racks should always be supported underneath when carried or transported. They are available in solid colors, or color combinations to simplify coding identification. Each individual rack holds four tubes. Supplied two racks per pack.