Anti-IgG Goat Antibody (AzureSpectra 700)

Supplier: Azure Biosystems
AC2128
10147-350EA 245.8 CAD
10147-350
Anti-IgG Goat Antibody (AzureSpectra 700)
Antibodies
Type: Immunoglobulin G
Antigen: IgG
Clonality:
Clone:
Conjugation: AzureSpectra 700
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR