Kevlar® Nomex® 2610 Seamless Heat Resistant Gloves, Wells Lamont
Supplier: Wells Lamont
2610S 2610M 2610L 2610XL
75836-504PK 576.62 CAD
75836-504 75836-506 75836-508 75836-510
Kevlar® Nomex® 2610 Seamless Heat Resistant Gloves, Wells Lamont
Gloves
Kevlar® Nomex® heat resistant gloves protect up to 260 °C (500 °F).
Kevlar® Nomex® heat resistant gloves feature an outer shell that provides excellent thermal properties, allowing wearer to handle hot objects. These gloves are fully lined with cotton for softness. Gloves are ambidextrous, knit, and they feature a continuous cuff.
Ordering information: These gloves are sold by the dozen pieces.