KIMBLE® KimCote® GL 45 Media Bottles, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
KC14395-100 KC14395-250 KC14395-500 KC14395-1000 KC14395-2000
89097-368CS 480.73 CAD
89097-368 89097-370 89097-372 89097-374 89097-376
KIMBLE® KimCote® GL 45 Media Bottles, DWK Life Sciences
Bottles Cell Culture Bottles
Ideal for general laboratory use including mixing, storing or transporting culture media, chemicals, or solvents.
  • 30 mm Int.Ø. opening
  • Blue polypropylene linerless GL 45 screw thread cap
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
Autoclavable KimCote® safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Blue caps and clear pour rings included.