96-Well Opaque Skirted PCR Plate, Chemglass
Supplier: Chemglass
CLS-3614-04B CLS-3614-04W
89492-954CS 715.95 CAD
89492-954 89492-956
96-Well Opaque Skirted PCR Plate, Chemglass
Microplates PCR Plates
Thin wall, opaque, 96-well PCR plates are designed for chemiluminescent and fluorescent procedures
  • All plates are autoclavable
  • Available in both black and white
  • An alphanumeric grid helps in sample identification
Precision molded to ensure well-to-well and plate-to-plate uniformity.
Certifications: Certified RNase-, DNase-, Pyrogen-, and DNA-free.
Packaging: 10 plates packed in tamper-proof resealable bags, 100 per case.