Spinner Flask, Water Jacketed, Bioprocess, Chemglass
Supplier: Chemglass
CLS-1430-J100 CLS-1430-J101 CLS-1430-J1L CLS-1430-J1L1 CLS-1430-J250 CLS-1430-J251 CLS-1430-J3L CLS-1430-J3L1 CLS-1430-J500 CLS-1430-J501 CLS-1430-J6L CLS-1430-J6L1
89492-876EA 1992.36 CAD
89492-876 89492-878 89492-880 89492-882 89492-884 89492-886 89492-888 89492-890 89492-892 89492-894 89492-896 89492-898
Spinner Flask, Water Jacketed, Bioprocess, Chemglass
Flasks Spinner Flasks
Water jacketed, flat bottomed spinner flasks consist of a water jacketed spinner flask and a microcarrier style impeller with glass shaft that extends through the cap and PTFE paddle.
  • Manufactured from Type 1 Class A borosilicate glass
Flasks are typically used with a circulating water bath. Jacket volumes are approximate and flasks are not graduated.
Certifications: Glass conforms to ASTM Type 1, Class A requirements.
Accessories information: Port assemblies are constructed of 316 l electro-polished stainless steel. These port assemblies fitMore Product Information