Cytotoxicity Control Reagent, BioAssay Systems
Supplier: BioAssay Systems
CTTX-050
75877-944EA 254.17 CAD
75877-944
Cytotoxicity Control Reagent, BioAssay Systems
Controls and Calibration Verification Materials
A cytotoxic control agent (Saponin) for the Cell Viability Assay Kits.
Useful as a cytotoxic control agent in the CellQuanti-Blue (CQBL-05K) and CellQuanti-MTT (CQMT-500) Cell Viability Assay Kits.
IC50 = 0.002 wt%
Shelf life: 12 months