Total Protein Assay Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
QTPR-01K QTPR-100
75878-194EA 521.92 CAD
75878-194 75878-196
Total Protein Assay Kit, BioAssay Systems
Assays Protein Assays
For quantitative determination of total protein
  • Fast and sensitive. Assay takes 10 min. Linear detection range of 5 - 200 mg/dL protein.
  • Robust and Reliable. Substances in urine have minimal interference (< 2%).
  • Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day.
  • Method: OD600nm (Pyrogallol Red)
  • Samples: Urine, cerebrospinal fluid etc
Total protein concentration can be determined by several methods. However, protein dye-binding procedures are simple, reliable, rapid, readily automated and are becoming increasingly popular in assessing renal and blood brain barrier functions. BioAssay Systems’ Total Protein Assay Kit is based on an improved pyrogallol red-molybdate protein dye-binding assay. The color intensity at 600nm is directly proportional to the total protein concentration in the sample. This single reagent can be used for the quantitative determination of total protein in urine (proteinuria) and cerebrospinal fluid (blood brain barrier).