Microsyringes, PerkinElmer
Supplier: PerkinElmer
09904822 09904823 09904941 09904943
CA97012-198EA 321.03 CAD
CA97012-198 CA97012-200 CA97012-202 CA97012-216
Microsyringes, PerkinElmer
Syringes Syringes for Chromatography
These microsyringes are used for accurate and precise liquid delivery
  • Hand fitted to assure maximum accuracy
  • Composed of glass barrels and precision stainless steel needles
  • Features a blunt tip, required for use with a rheodyne injector