HyClone™ Fetal Bovine Serum Characterized (United States)

Supplier: Cytiva
SH30071.03 SH30071.01HI SH30071.02HI SH30071.03 3007103IR30-45 SH30071.03HI
16777-014EA 1460.2 CAD
16777-014 CA76237-676 CA76237-678 CA16777-010 CA82013-572 CA16777-014 CA76324-922 CA83007-198 CA76324-864 CA76324-924 CA76324-862
HyClone™ Fetal Bovine Serum Characterized (United States)
Sera (Serum)
Enhance growth of cells with U.S.sourced Cytiva HyClone™ Characterized Fetal Bovine Serum (FBS). More economical than Defined FBS, but meets the requirements of most discriminating cell culturists.

Tested for endotoxin, hemoglobin, and additional biochemical assays and filtered through serial 100 nm pore-size rated filters.
Order Now

SPECIFICATIONS

Environmentally Preferable
Concentration Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl
Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl
Endotoxin: ≤25 EU/ml
Hemoglobin: ≤25 mg/dl
Endotoxin: ≤5 EU/ml
Hemoglobin: ≤15 mg/dl
Origin US US US US
Endotoxin ≤25 EU/ml ≤25 EU/ml ≤25 EU/ml ≤5 EU/ml
Hemoglobin <25 mg/dl <25 mg/dl <25 mg/dl ≤15 mg/dl
Storage temperature Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C. Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C. Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C. Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR