Adventurer® AXW Balances, Ohaus®

Supplier: Ohaus
AX413/W AX513/W AX3102/W AX5102/W AX214/W
75871-490EA 5488.59 CAD
75871-490 75871-492 75871-494 75871-496 75871-622
Adventurer® AXW Balances, Ohaus®
Balances and Scales
OHAUS® Adventurer® AXW incorporates all of the applications necessary for routine weighing and measurement activities.

  • Stability, accuracy, and fast operation
  • Color touchscreen and connectivity options
  • Space-saving draftshield

Stability, accuracy, and fast operation ensure optimal weighing results - This trio of characteristics ensures the most important aspects of routine laboratory weighing are covered: Accurate results, achieved quickly, that you can count on.

Color touchscreen and connectivity options power a modern weighing experience - The wide viewing angle color touchscreen offers quick access to all of Adventurer® W’s applications. Dual USB ports and the ability to store GLP/GMP information assist in the monitoring and reporting of data.

Space-saving Draftshield Improves User Experience and Accessibility — The two piece, top-mounted draftshield doors open wide on both sides, providing ample room for sample placement, and also reduce the footprint of the draftshield in the rear of the balance when open.

Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS® Adventurer® W incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, GLP/GMP compliance capabilities, two USB ports, ingenious draftshield, and much more, Adventurer® W is the most complete balance in its class.

Certifications: Product Safety: CSA C22.2 No. 61010-1, UL 61010-1, IEC 61010-1. Electromagnetic Compatibility: FCC Part 15 Class A, ICES-001 Class A, IEC 61326-1.

Ordering information: Manufacturer’s two-year parts and labor warranty (U.S. and Canada).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR