Oil for Welch® Vacuum Pumps, Chemglass
Supplier: Chemglass
AF-0360-10 AF-0360-11 AF-0360-12 AF-0360-13 AF-0360-14 AF-0360-16 AF-0360-17
89028-178EA 97.02 CAD
89028-178 89028-180 89028-182 89028-184 89028-186 89028-190 89028-192
Oil for Welch® Vacuum Pumps, Chemglass
Lubricants
Oil is specially formulated for use in either belt driven or direct drive vacuum pumps (AF-0349 or AF-0350)
  • Oil is available in three types, Duo-Seal®, Premium and Gold
The Duo-Seal oil is for use with belt driven (low RPM) AF-0350 pumps.
The Premium oil is a triple distilled hydrocarbon oil resistant to breakdown at the high RPM of direct drive pumps.
The Gold oil is a double distilled synthetic hydrocarbon oil that has no aromatic compounds or sulfur. Gold oil will offer longer life and superior protection for the internal components in corrosive pumping applications.