Welch® Belt Driven Vacuum Pumps, Chemglass
Supplier: Chemglass
AF-0350-01 AF-0350-02 AF-0350-03
80060-096EA 11676.63 CAD
80060-096 80060-098 80060-100
Welch® Belt Driven Vacuum Pumps, Chemglass
Pumps Vacuum Pumps
This general purpose two stage belt driven pumps are designed to operate at lower RPM allowing them to run cooler and reach an ultimate vacuum of 1 x 10-4 Torr.
  • Lower speed operation reduces the wear on the pump components ensuring a longer life
  • All pumps are equipped with a gas ballast
This pump is ideally suited for freeze drying, concentrating, Schlenk line, degassing, vacuum distillations, vacuum drying.