Vacuum Manifold, Inert Line, Schlenk, Chemglass
Supplier: Chemglass
AF-0452-01 AF-0452-02 AF-0452-03 AF-0452-04
80064-434EA 5877.63 CAD
80064-434 80064-436 80064-438 80064-440
Vacuum Manifold, Inert Line, Schlenk, Chemglass
Manifolds
Vacuum manifolds constructed using 4 mm double oblique hollow high vacuum with greased stopcocks on all four ports.
Ports are fitted with two 14/20 inner joints and two 24/40 inner joints and are spaced 5" center to center. This configuration permits use directly with vessels having these joint sizes. All four ends of the manifold have No. 15 O-ring joints permitting the trap assembly to be attached to either the right or left side of the vacuum bank.
Trap assembly is a rugged two piece design with 0 to 12 mm Chem-Vac™ Chem-Cap® valve as the shut off. Stopper and body connection is made via a 57 mm flat O-ring flange.
Ordering information: Complete system is supplied with five SS pinch clamp No. 28, five Viton O-rings No. 116. Manifold is supplied with two O-ring joint airfree stoppers No.15, three Style "B" airfree adapters for 15 mm O-ring joint to 10 mm serrated hose connection. Trap assembly is supplied complete with a horseshoe style clamp 57 mm flange and Viton o-ring No. 331. Trap is supplied complete with one Style "A" airfree adapter for No. 15 O-ring joint to 3/4" OD straight tubing adapter and one airfree adapter No.15 O-ring joint to 3/4" bent tubing for connection to heavy wall flexible vacuum tubing.