LaboPlast®/SteriPlast® Spoon Spatula, Bürkle
Supplier: BURKLE INC
5378-0031 5378-1031
75876-074PK 76.88 CAD
75876-074 75876-076
LaboPlast®/SteriPlast® Spoon Spatula, Bürkle
Spatulas
The spoon spatula is a double spatula with a 0.5 ml (0.02 oz.) spoon and a 17 mm (0.67") spatula blade.
  • Polystyrene construction
  • Production, assembly, and packaging according to Cleanroom Class 7
  • Complies with EU food requirements and FDA regulations
  • Optionally packaged individually and sterilized by gamma rays
The spoon spatula has many different uses in lab applications. It may be used as a sampling spoon, for measuring or transferring small amounts, as a scraper, or for mixing and stirring. Specially designed for disposable use, it is resistant toMore Product Information