Volumetric Pipettes, Class A, Glass, Hach
Supplier: Hach
1451540 1451534 1451536 1451541 1451537
CA14515-40EA 76.39 CAD
CA14515-40 CA11014-154 CA11014-156 CA11014-354 CA11028-076
Volumetric Pipettes, Class A, Glass, Hach
Pipettes Graduated Pipettes
Made from Class A glass, pipettes are calibrated 'to deliver'.
  • Unserialized
  • Color-coded
Certifications: ASTM E969 specification for accuracy and ASTM E1273 for color coding.