Plate Count Agar, Teknova
Supplier: TEKNOVA CA
P0100 P0101
CA100216-642PK 841.81 CAD
CA100216-642 CA100218-912
Plate Count Agar, Teknova
Media for Microbiology Prepared Media for Microbiology
Plate count agar are a standard method medium used for the enumeration of bacteria in water, waste water, dairy products and foods.