Anti-CD24 Rat Monoclonal Antibody (violetFluor® 450) [clone: M1/69]

Supplier: Tonbo Biosciences
75-0242-U025 75-0242-U100
75875-726EA 139.04 CAD
75875-726 75875-728
Anti-CD24 Rat Monoclonal Antibody (violetFluor® 450) [clone: M1/69]
Antibodies
The M1/69 antibody reacts with mouse CD24, a cell surface molecule also known as Heat Stable Antigen (HSA). CD24 is a GPI-linked glycoprotein involved in cell adhesion and is a ligand for CD62P (P-selectin). It is expressed on erthrocytes, thymocytes, peripheral lymphocytes and myeloid lineage cells. The expression of CD24 is used to delineate stages of of lymphocyte development in the bone marrow.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: M1/69
Conjugation: violetFluor® 450
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR