Anti-CD5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53-7.3]

Supplier: Tonbo Biosciences
50-0051-U025 50-0051-U100
75875-712EA 56.88 CAD
75875-712 75875-714
Anti-CD5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53-7.3]
Antibodies
The 53-7.3 monoclonal antibody is specific for mouse CD5, a 67 kDa protein that is expressed on most thymocytes, mature T cells and the B-1 subset of B cells. CD5 is a member of the scavenger receptor cysteine-rich protein superfamily and is a ligand for the c-type lectin CD72. CD5 is also known as Leu-1 and functions to modulate antigen receptor signaling on T and B cells.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR