Bottles, Wide-Mouth, with Cap, Clear, Type III Soda-Lime Glass, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
W217970 W217971 W217972 W217973 W217925 W217926 W217927 W217928 W217921 W217922 W217924
10847-508CS 213.38 CAD
10847-508 10847-510 10847-512 10847-514 10847-582 10847-584 10847-586 10847-588 10847-672 10847-678 10847-682
Bottles, Wide-Mouth, with Cap, Clear, Type III Soda-Lime Glass, Wheaton
Bottles Glass Bottles
Clear, wide mouth, volumetric glass bottles are made of Type III soda-lime glass.
  • Supplied with attached screw caps
  • Ideal for field or general laboratory use