PRO Internal Baths with Base Controller, Lauda-Brinkmann
Supplier: Lauda-Brinkmann
L000306 L000307 L000308 L000309 L000310 L000311 L000546 L000547 L000548 L000450 L000451
75840-804EA 29383.59 CAD
75840-804 75840-806 75840-808 75840-810 75840-812 75840-814 75840-834 75840-836 75840-838 75840-848 75840-850
PRO Internal Baths with Base Controller, Lauda-Brinkmann
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
PRO units have advantages such as low filling volume combined with powerful Varioflex pump, resulting in rapid temperature change. Environment Friendly (GWP).
  • Bright high contrast OLED display
  • Programmer with 1 program, 20 segments
  • Removable remote control
  • Temperature display in °C or °F
  • Menu navigation in 5 languages
LAUDA PRO Thermostats are optimized for interal bath applications and offer excellent temperature stability
The BASE Controller features a bright OLED display that is removable from the unit, allowing for flexible operation.