SP Wilmad-LabGlass Distilling Heads with Vacuum Port, SP Industries
Supplier: SP Industries
LG-6115-126 ML-870-702 LG-6115-100
14209-102EA 1064.65 CAD
14209-102 14205-244 14208-972
SP Wilmad-LabGlass Distilling Heads with Vacuum Port, SP Industries
Distillation Components Distillation Heads
These distilling heads have a one piece design with the condenser angled at 75° and a vacuum port on product side which can be left open for atmospheric distillations.
  • Borosilicate glass
A Standard Taper 10/30 outer joint is provided for a 75mm immersion thermometer. See thermometer adapters (LG-1453) for use with models ML-870.