Dextrose monohydrate, powder USP
Supplier: Spectrum Chemical Mfg. Corp.
DE145-100GM
DE145-45KGBL
75810-996EA
184.26
CAD
75810-996
75810-998
Dextrose monohydrate, powder USP
Dextrose monohydrate
Dextrose, Monohydrate, Powder, USP - All Spectrum Chemical USP products, including all USP dextrose products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Synonyms: Corn Sugar; D (+)-Glucopyranose; Glucose; D-Glucose.
Storage Temperature:
Ambient |
CAS Number:
5996-10-1 |
Specification Test Results
Assay (anhydrous basis) | 97.5 -102.0% |
Water determination | 7.5 - 9.5% |
Related substances | To pass test |
Color and clarity of solution | To pass test |
Conductivity @ 25°C | Max 20 uS/cm |
Elemental impurities | As reported |
Dextrin | To pass test |
Soluble starch, sulfites | To pass test |
Identification | To pass test |
Certified Halal | — |
Residual solvents | To pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...