Avantor® Alltima HP C18 Hi-Load, HPLC Columns
Supplier: VWR
Alltima®
87686 22185 87688 87700 87610 87687 22174 87690 5135351 22183 87624/N 87692 87694 87612 87827 AHP-3C18H-250D 22195 22194 22173 87697 87698 87506 22175 22176 22231 22193 87685 87611 22184 AHP-5C18H-50DM 22230 87701/N 87699
75830-806EA 943.7 CAD
75830-806 75797-502 75797-524 75797-504 75797-526 75797-506 75797-508 76398-380 75797-494 75797-472 75797-474 75797-476 75797-530 75797-478 75797-510 75797-532 75798-908 75797-490 75797-492 75830-818 76398-376 75797-534 75797-512 75797-514 75797-518 75797-482 75798-912 75797-484 75797-520 75798-910 75797-486 75797-488 75797-480
Avantor® Alltima HP C18 Hi-Load, HPLC Columns
Chromatography Columns
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica.
  • Excellent column stability
  • Low to no detectable column bleed
  • Multiple selectivity options
These columns have highest carbon load for superior retention and loadability for high resolution of complex samples.
These product family combines the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
Column hardware: Stainless steel.