Avantor® Alltima HP, HILIC HPLC Columns
Supplier: Avantor
86461 86463 86462 86465 86464 86466 86479/N 86480/N
75797-600EA 1009.64 CAD
75797-600 75797-602 75797-604 75797-606 75797-608 75797-610 75797-612 75797-614 75797-616 75797-618 75797-620 75797-622 75797-624 75797-626 75797-628 75797-630 75797-632 75797-634 75797-636 75797-638 75797-640 75797-642 75797-644 75797-646 75797-648 75797-652 75797-656 75797-658 75830-800 75830-802 76398-338
Avantor® Alltima HP, HILIC HPLC Columns
Chromatography Columns
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica. The Avantor® Alltima HP product family combines the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
  • Excellent column stability
  • Low to no detectable column bleed
  • Multiple selectivity options
Hydrophilic interaction chromatography uses small amounts of water for increased sensitivity with microbore applications.
Very polar analytes thatMore Product Information