Avantor® Alltima HP C18 EPS, HPLC Columns
Supplier: HICHROM LIMITED
87716 87706 87703 87704
75798-898EA 1838.52 CAD
75798-898 75798-900 75799-168 75799-170 75799-172 75799-174 75799-176 75799-178 75799-180 75799-182 75799-184 75799-188 75799-190 75830-822 76398-340
Avantor® Alltima HP C18 EPS, HPLC Columns
Chromatography Columns
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica.
  • Excellent column stability
  • Low to no detectable column bleed
  • Multiple selectivity options
Avantor® Alltima HP C18 EPS - Greater retention and enhanced peak symmetry for polar compounds for reversed-phase applications where C18 is too retentive (USP Code: L1).
The Avantor® Alltima HP product family combines the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.