Avantor® Alltima HP C18 AQ, HPLC Columns
Supplier: Avantor
87816 87832 87807 87808 22538 87821 87819 87820 87824/N
75797-464EA 1765.4 CAD
75797-464 75797-466 75797-468 75797-470 75798-858 75798-860 75798-862 75798-864 75798-866 75798-868 75798-870 75798-872 75798-874 75798-876 75798-878 75798-880 75798-882 75798-884 75798-886 75798-888 75798-890 75798-892 75798-894 75798-896 75799-154 75799-158 75799-160 75799-164 75799-166 75830-840 75830-842 76398-344
Avantor® Alltima HP C18 AQ, HPLC Columns
Chromatography Columns
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica.
  • Excellent column stability
  • Low to no detectable column bleed
  • Multiple selectivity options
Avantor® Alltima HP C18 AQ - 100% wter wettable for applications requiring high aqueous mobile phases (USP code: L1).
The Avantor® Alltima HP product family combines the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.