SpectraTube™ Centrifuge Tubes, MTC Bio
Supplier: MTC Bio
C2750_WARDS C2751_WARDS C2716_WARDS C2715_WARDS
470224-998CS 510.85 CAD
470224-998 470225-000 470225-002 470225-004
SpectraTube™ Centrifuge Tubes, MTC Bio
Tubes Centrifuge Tubes
A new twist on a staple in the life science laboratory.
  • Uniform wall thickness
  • Store samples at temperatures as low as –86 °C
  • Color-coded caps for easy sample identification
  • Sterilized by ethylene oxide
  • RNAse-, DNAse-, pyrogen-, and cytotoxin-free
Available in Red, Orange, Yellow, Green, and Blue, these tubes are packaged in either styrofoam racks or in bulk bags. Racked tubes are available for purchase by individual rack, or by the case. Bulk bags of tubes are available for purchase by the case.
Perfect for any lab conducting multiple studies, day of the week studies, or any experiments requiring multiple sample types or solutions. All five cap varieties are flat, brightly colored, and frosted to make them easy to write on. The tubes have uniform wall thickness for clarity and centrifugal strength.
Caution: This product is designed for educational and teaching laboratories and no certificate of analysis is available.