Vacuum Drying Oven, Yamato
Supplier: Yamato Scientific
ADP-200C ADP-210C ADP-300C ADP-310C
75846-682EA 6319.56 CAD
75846-682 75846-684 75846-686 75846-688
Vacuum Drying Oven, Yamato
Ovens Vacuum Ovens
This small benchtop oven uses a decompressed direct heating chamber for vacuum drying.
  • Operating Pressure Range: 101 to ~0.1 kPa (760 to ~1 Torr)
  • Stainless Steel Material
  • Bourdon Tube-Type Vacuum Gauge
  • Mica Heater
  • Temperature Control Accuracy: ±1.5 °C (at 240 °C)
The user-friendly oven is easy to maintain and has straight-forward input of parameters settings and customizable communication ports
The digital PID controller supports fixed temperature, quick auto stop, auto stop and start, calibration offset, and program operations. Each circuitMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Insulation Rock Wool
Sensor K Thermocouple
Timer 1 Minute to 99 Hours, 59 Minutes. and 100 Hours to 999 Hours and 50 Minutes