Avantor Single-Use Tank Liners, Medical Grade, Avantor Fluid Handling
Supplier: AVANTOR FLUID HANDLING, LLC
1003001 1003003 1003004 1003005 1003006 1003007 1003008 1003009 1003010 1003011 1003012
75874-384CS 971.02 CAD
75874-384 75874-388 75874-390 75874-392 75874-394 75874-396 75874-398 75874-400 75874-402 75874-404 75874-406
Avantor Single-Use Tank Liners, Medical Grade, Avantor Fluid Handling
Bags Media Bags
These tank liners are single-use bags with an open top, designed to fit in cylindrical vessels.
  • No cleaning after media preparation
  • Medical grade, multi-layer film
  • For bioprocess applications
These bags are used as a liner into a vessel to avoid cleaning of the vessel after media and buffer preparation.
Tank Liners are made of a clean medical grade multi-layer film designed for bioprocess applications. The fluid contact layer is a medical grade LLDPE. To minimize gas diffusion, an EVOH layer is coextruded between the inner and outer layers. The outer, non-contact strength layer is formed from polyamide and is coextruded with polyethylene to create a bonding layer.
Tank liners are available in both 2D and 3D style. Tank liners will be delivered gamma irradiated and ready-to-use.
After installing the tank liner into a vessel and filling it with liquid, powder can be added to the vessel and the mixing process can start by using an overhead agitator. When mixing is finished, it’s possible to pump the mixed liquid into sterile bags by using a sterile filter in order to have sterile media and buffers.
Certifications: USP Class VI and animal derived component free (ADCF).
Ordering information: VWR® is also able to deliver customer-specific tank liners. The bags can be made for almost all vessels up to 1.500 L, optionally bottom drain. For more information about customization, please visit vwr.com/single-use.