VeriSpec™ Tartrate - OOC(CHOH)₂COO₂₋ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010945-100C RV010945-250C RV010945-500C
RCRV010945100CEA 188.02 CAD
RCRV010945100C RCRV010945250C RCRV010945500C
VeriSpec™ Tartrate - OOC(CHOH)₂COO₂₋ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards Anionic Standards Tartrate Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility