VeriSpec™ Sulfate - SO₄²⁻ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010943-100N RV010943-250N RV010943-500N
RCRV010943100NEA 173.8 CAD
RCRV010943100N RCRV010943250N RCRV010943500N
VeriSpec™ Sulfate - SO₄²⁻ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards Anionic Standards Sulphate Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility