VeriSpec® Strontium - Sr²⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010941-100N RV010941-250N RV010941-500N
RCRV010941100NEA 165.9 CAD
RCRV010941100N RCRV010941250N RCRV010941500N
VeriSpec® Strontium - Sr²⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards Cationic Standards Strontium Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility