VeriSpec® Silicate - SiO₃²⁻ Standard for Ion Chromatography 1.000 +/- 0.002 g/L in NaOH 1%, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010939-250N RV010939-500N RV010939-100N
RCRV010939500NEA 369.23 CAD
RCRV010939500N RCRV010939100N RCRV010939250N
VeriSpec® Silicate - SiO₃²⁻ Standard for Ion Chromatography 1.000 +/- 0.002 g/L in NaOH 1%, RICCA Chemical Company
Standards Anionic Standards Silicate Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility