VeriSpec™ Potassium - K⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010937-100N
RV010937-250N
RV010937-500N
RCRV010937100NEA
173.8
CAD
RCRV010937100N
RCRV010937250N
RCRV010937500N
VeriSpec™ Potassium - K⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards
Cationic Standards
Potassium Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility