VeriSpec® Phosphate - PO₄³⁻ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010936-100N RV010936-250N RV010936-500N
RCRV010936100NEA 172.22 CAD
RCRV010936100N RCRV010936250N RCRV010936500N
VeriSpec® Phosphate - PO₄³⁻ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards Anionic Standards Phosphate Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility