VeriSpec™ Magnesium - Mg²⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010923-100N RV010923-250N RV010923-500N
RCRV010923100NEA 158 CAD
RCRV010923100N RCRV010923250N RCRV010923500N
VeriSpec™ Magnesium - Mg²⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards Cationic Standards Magnesium Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility