VeriSpec™ Formate - HCOO⁻ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010916-100N RV010916-500N RV010916-250N
RCRV010916500NEA 374.51 CAD
RCRV010916500N RCRV010916100N RCRV010916250N
VeriSpec™ Formate - HCOO⁻ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards Anionic Standards Formate Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility